missing translation for 'onlineSavingsMsg'
Learn More

Lanthanum rod, 12.7mm (0.5in) dia, 99.9% (metals basis excluding Ta), Thermo Scientific Chemicals

110.55 € - 591.95 €

Chemical Identifiers

CAS 7439-91-0
Molecular Formula La
Molecular Weight (g/mol) 138.91
MDL Number MFCD00011066
InChI Key FZLIPJUXYLNCLC-UHFFFAOYSA-N
Synonym in chloride, ion 2, lanthanum, ion la2, atom, ingot, pieces, lanthanum, pieces, 2+ ion, foil, 3n, ii cation
PubChem CID 23926
ChEBI CHEBI:33336
IUPAC Name lanthanum
SMILES [La]
View More Specs

Products 4
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11383828
View Documents
Thermo Scientific Alfa Aesar
040293.HF
10 mm
110.55 €
10mm
Please to purchase this item. Need a web account? Register with us today!
11353828
View Documents
Thermo Scientific Alfa Aesar
040293.DH
25 mm
216.08 €
25mm
Please to purchase this item. Need a web account? Register with us today!
11373828
View Documents
Thermo Scientific Alfa Aesar
040293.DJ
50 mm
337.68 €
50mm
Please to purchase this item. Need a web account? Register with us today!
11363828
View Documents
Thermo Scientific Alfa Aesar
040293.DM
100 mm
591.95 €
100mm
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under argon
Specifications

Chemical Identifiers

7439-91-0
138.91
FZLIPJUXYLNCLC-UHFFFAOYSA-N
23926
lanthanum
La
MFCD00011066
in chloride, ion 2, lanthanum, ion la2, atom, ingot, pieces, lanthanum, pieces, 2+ ion, foil, 3n, ii cation
CHEBI:33336
[La]

Specifications

Silver to Gray
La
UN3208
FZLIPJUXYLNCLC-UHFFFAOYSA-N
lanthanum
23926
99.9%
≈0.80g/mm
(metals basis excluding Ta)
7439-91-0
MFCD00011066
in chloride, ion 2, lanthanum, ion la2, atom, ingot, pieces, lanthanum, pieces, 2+ ion, foil, 3n, ii cation
[La]
138.91
CHEBI:33336
Mylar under Argon
Cast Rod
Lanthanum rod, 12.7mm (0.5 in.) dia.
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H228-H261
Flammable solid.
In contact with water releases flammable gas.

P210-P240-P241-P280-P370+P378q

H228

missing translation for 'dotInformation' : Transport Hazard Class: 4.3; Packing Group: II; Proper Shipping Name: METALLIC SUBSTANCE, WATER-REACTIVE, N.O.S.

missing translation for 'einecsNumber' : 231-099-0

missing translation for 'tsca' : Yes

missing translation for 'storageNote' :

  • Packaged under argon

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.