Learn More
Nitritex™ BioClean Legacy™ Latex Cleanroom Gloves
Used for clean room work. Nitritex™ BioClean Legacy™ Latex Cleanroom Gloves are natural colored and provide unbeatable comfort.
Brand:  Nitritex™ BLA202-M
 View more versions of this product
Product Code. 12705825
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
  • Particulates: Processed in a cleanroom environment to ensure compatibility with ISO Class 4 operating conditions.
  • Physical Properties: Complies with the requirements of the Glove Standard EN420:2003, the European Medical Gloves Standard EN455 (Parts 1, 2 & 3) & EN374-1, 2 & 3:2003.
  • Sterile: Non-Sterile
  • Quality Systems: Manufactured in a facility holding the ISO 9001:2008 and 89/686/EEC article 11b certifications. Processed in an NEBB certified ISO Class 4 cleanroom environment.
  • Sampling & Inspection: In accordance with ISO 2859.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
ISO 9001:2015 and (EU)2016/425, EN420:2003 + A1:2009, EN374-1:20016, EN374-5:2016 and EN421:2010, EN455-2:2015, (EC)1935/2004 and (EC)2023/2006
Natural
Yes
Latex
Cleanroom Gloves
Non-sterile
100 pieces per sealed inner PE bag; one inner PE bag per sealed outer PE bag; 10 outer bags per lined carton (1000 pieces)
Category 3 - Complex Design Personal Protective Equipment conforming to PPE Directive 89/686/EEC
Single-use
No
Powder Free
Medium
Beaded/Rolled
300 mm