missing translation for 'onlineSavingsMsg'
Learn More

Nitritex™ BioClean Legacy™ Latex Cleanroom Gloves
BIOPHARMA

Used for clean room work. Nitritex™ BioClean Legacy™ Latex Cleanroom Gloves are natural colored and provide unbeatable comfort.

23.60 € - 339.00 €

Specifications

Classification Category 3 - Complex Design Personal Protective Equipment conforming to PPE Directive 89/686/EEC
Color Natural
Disposable Single-use
Laboratory/Cleanroom Use Yes
Latex Free No
View More Specs

Products 9
Product Code Brand Size Price Quantity & Availability  
Product Code Brand Size Price Quantity & Availability  
12795815
View Documents
Nitritex™
BLA201-S
Small This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
12715825
View Documents
Nitritex™
BLA203-L
Large This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15208188
View Documents
Nitritex™
BLA203-L
Large
339.00 €
Pack of 1000
In Stock
to see stock.
Add to basket
 
15288178
View Documents
Nitritex™
BLA201-S
Small
339.00 €
Pack of 1000
In Stock
to see stock.
Edge
Add to basket
 
15186753
View Documents
Nitritex™
BLA205-XXL
2X-Large
339.00 €
Pack of 1000
Estimated Shipment: 29-04-2025
to see stock.
Add to basket
 
15298178
View Documents
Nitritex™
BLA202-M
Medium
339.00 €
Pack of 1000
Estimated Shipment: 10-07-2025
to see stock.
Add to basket
 
12705825
View Documents
Nitritex™
BLA202-M
Medium This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15278178
BIOPHARMA
View Documents
Nitritex™
BLA200-XS
X-Small
339.00 €
Pack of 1000
Estimated Shipment: 29-04-2025
to see stock.
Add to basket
 
12725825
View Documents
Nitritex™
BLA204-XL
X-Large This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

  • Particulates: Processed in a cleanroom environment to ensure compatibility with ISO Class 4 operating conditions.
  • Physical Properties: Complies with the requirements of the Glove Standard EN420:2003, the European Medical Gloves Standard EN455 (Parts 1, 2 & 3) & EN374-1, 2 & 3:2003.
  • Sterile: Non-Sterile
  • Quality Systems: Manufactured in a facility holding the ISO 9001:2008 and 89/686/EEC article 11b certifications. Processed in an NEBB certified ISO Class 4 cleanroom environment.
  • Sampling & Inspection: In accordance with ISO 2859.
Specifications

Specifications

Category 3 - Complex Design Personal Protective Equipment conforming to PPE Directive 89/686/EEC
Single-use
No
Powder Free
Non-sterile
300 mm
Natural
Yes
Latex
Cleanroom Gloves
Beaded/Rolled
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Nitritex™ BioClean Legacy™ Latex Cleanroom Gloves

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.