missing translation for 'onlineSavingsMsg'
Learn More

Fisher BioReagents™ LB Broth, Miller (Granulated)

Used as a microbial growth medium for the cultivation of E. coli strains for the preparation of plasmid DNA and recombinant proteins.

Brand:  Fisher BioReagents™ BP9723-2

 View more versions of this product

Product Code. 11325992

  • 381.00 € / 2kg
Estimated Shipment: 21-11-2024
to see stock.
Edge
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Directions for Media Preparation: Add 25g to 1L of purified water, such as BP2820 (Fisher BioReagents Microbial Cell Culture Grade Water) then adjust pH to 7.2. Heat and frequently agitate to completely dissolve. Before use, sterilize by autoclaving for 15 minutes.
  • Sodium chloride concentration of Miller formulation is 2X the concentration of the Lennox formulation to allow for optimal salt concentration for a specific strain
  • Format: Granulated powder
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Dehydrated
Bacterial Culture
Microbiology Media
2000 g
LB Broth
Casein Peptone 10g/L, Yeast extract 5g/L, Sodium Chloride 10g/L
Poly Bottle
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers