missing translation for 'onlineSavingsMsg'
Learn More

Fisher BioReagents™ LB Broth, Miller (Granulated)

Used as a microbial growth medium for the cultivation of E. coli strains for the preparation of plasmid DNA and recombinant proteins.

381.00 €

Specifications

Media Type LB Broth
Formulation Casein Peptone 10g/L, Yeast extract 5g/L, Sodium Chloride 10g/L
View More Specs

Products 1
Product Code Brand Quantity Packaging Type Price Quantity & Availability  
Product Code Brand Quantity Packaging Type Price Quantity & Availability  
11325992
View Documents
Fisher BioReagents™
BP9723-2
2000 g Poly Bottle
381.00 €
2kg
Estimated Shipment: 03-12-2024
to see stock.
Edge
Add to basket
 
Description

Description

  • Directions for Media Preparation: Add 25g to 1L of purified water, such as BP2820 (Fisher BioReagents Microbial Cell Culture Grade Water) then adjust pH to 7.2. Heat and frequently agitate to completely dissolve. Before use, sterilize by autoclaving for 15 minutes.
  • Sodium chloride concentration of Miller formulation is 2X the concentration of the Lennox formulation to allow for optimal salt concentration for a specific strain
  • Format: Granulated powder
Specifications

Specifications

LB Broth
Casein Peptone 10g/L, Yeast extract 5g/L, Sodium Chloride 10g/L
Videos
SDS
Documents

Documents

Product Certifications