Learn More
Lovibond™ LC 100 Hand-Held Imaging Spectrophotometer
Creates conformance reports in either PDF or Excel formats that can easily be shared with PC or compatible printer
Brand:  Lovibond™ 401100
 View more versions of this product
Product Code. 15564283
  • 2525.00 € / Each
Explore more special offers
Description
Description
  • Each measurement combines 27 images illuminated with different colours and from different directions
  • Enables preview of measurement area and storage and recall of image after measurement
  • Measures and stores up to 20 reference standards with each date and time stamped with option to name
  • Stores up to 350 sample measurements on device and compares to reference standards
  • Samples can be tagged via on-screen keyboard for text or via built in voice recorder
  • For indoor use only
LC 100 and SV 100 Integrated Kit
  • Contents: spectrophotometer, holder, sample cells, USB cord, carrying case
  • Food and Beverages, Textile, Chemicals, Pharmaceutical and Cosmetics, Medical and Clinical
Specifications
Specifications
TFT
Food and beverages, coatings, household products, pharmaceuticals and cosmetics, textiles, chemicals, medical and clinical, Plastics
Spectrophotometer, holder, sample cells, USB cord, carrying case
1.8 sec.