Learn More
Lovibond™ LC 100 Hand-Held Imaging Spectrophotometer
Creates conformance reports in either PDF or Excel formats that can easily be shared with PC or compatible printer
2525.00 €
Specifications
Display | TFT |
---|---|
Includes | Spectrophotometer, holder, sample cells, USB cord, carrying case |
Measurement Time | 1.8 sec. |
Product Code | Brand | For Use With (Application) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | For Use With (Application) | Price | Quantity & Availability | |||||
15564283
|
Lovibond™
401100 |
Food and beverages, coatings, household products, pharmaceuticals and cosmetics, textiles, chemicals, medical and clinical, Plastics |
2525.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- Each measurement combines 27 images illuminated with different colours and from different directions
- Enables preview of measurement area and storage and recall of image after measurement
- Measures and stores up to 20 reference standards with each date and time stamped with option to name
- Stores up to 350 sample measurements on device and compares to reference standards
- Samples can be tagged via on-screen keyboard for text or via built in voice recorder
- For indoor use only
LC 100 and SV 100 Integrated Kit
- Contents: spectrophotometer, holder, sample cells, USB cord, carrying case
Food and Beverages, Textile, Chemicals, Pharmaceutical and Cosmetics, Medical and Clinical
Specifications
TFT | |
Spectrophotometer, holder, sample cells, USB cord, carrying case | |
1.8 sec. |