missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Macherey-Nagel™ TLC Spray Visualization Reagents
Used for postchromatographic detection reactions in thin-layer chromatography. Macherey-Nagel™ TLC Spray Visualization Reagents are a selection of frequently used spray reagents that are suited for spraying or dipping TLC plates. Glass laboratory sprayer (mfr. no. 814101) sold separately.
Brand: Macherey-Nagel™ 814919
This item is not returnable.
View return policy
Description
- Broad selection of premium reagents available for detection of a wide range of substances
- Can be used for spraying or dipping of TLC plates
- Packaged in 100mL screw-top amber bottles
- Use with chemically-resistant sprayers, including Macherey-Nagel glass laboratory sprayer with rubber bulb (mfr. no. 814101, sold separately)
Specifications
7705-08-0 | |
0.79g/cm3 | |
Aniline phthalate spray reagent | |
Aniline phthalate in 2-Propanol–Ethanol (1:1) | |
TLC plates | |
C6H7N—C6H6O4 | |
Amber bottle with screw cap | |
100 mL |
Colorless | |
Spraying or Dipping | |
0.2 to <1% Aniline; 1 to <10% Phthalic Acid | |
Detection of reducing sugars, oxohalic acids | |
2-Propanol–Ethanol (1:1) | |
Alcoholic | |
Spray Visualization Reagent | |
Solution |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Supplemental information
- Aniline phthalate spray reagent, 100 mL
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 3
- Specific target organ toxicity after repeated exposure Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H226-Flammable liquid and vapour.
- H319-Causes serious eye irritation.
- H336-May cause drowsiness or dizziness.
- H373-May cause damage to organs.
Supplemental information
- MIXTURE LIST-Contains : aniline, ethanol, 2-propanol, phthalic acid