missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ TLC Spray Visualization Reagents

Used for postchromatographic detection reactions in thin-layer chromatography. Macherey-Nagel™ TLC Spray Visualization Reagents are a selection of frequently used spray reagents that are suited for spraying or dipping TLC plates. Glass laboratory sprayer (mfr. no. 814101) sold separately.

41.45 €

Specifications

CAS 7705-08-0
Detection Method Spraying or Dipping
For Use With (Equipment) TLC plates
Packaging Type Amber bottle with screw cap
Product Type Spray Visualization Reagent
View More Specs

Products 12
Product Code Brand Description Matrix For Use With (Application) Price Quantity & Availability  
Product Code Brand Description Matrix For Use With (Application) Price Quantity & Availability  
10669191
View Documents
Macherey-Nagel™
814203
Ninhydrin spray reagent Ethanol Detection of amino acids, amines, amino sugars
41.45 €
100mL
Estimated Shipment: 28-02-2025
to see stock.
Add to basket
 
10565371
View Documents
Macherey-Nagel™
814920
Bromocresol green spray reagent 2-Propanol Detection of organic acids
41.45 €
100mL
Estimated Shipment: 21-03-2025
to see stock.
Add to basket
 
10609581
View Documents
Macherey-Nagel™
814302
Molydatophosphoric acid spray reagent Ethanol Detection of lipids, sterols, steroids, reducing compounds
41.45 €
100mL
Estimated Shipment: 04-03-2025
to see stock.
Add to basket
 
10164991
View Documents
Macherey-Nagel™
814402
Dragendorff-Munier spray reagent Water Detection of alkaloids and other nitrogen compounds
41.45 €
100mL
Estimated Shipment: 20-02-2025
to see stock.
Add to basket
 
10650152
View Documents
Macherey-Nagel™
814404
Potassium Hexacyanoferrate(III) spray reagent Water Detection of acetylsalicylic acid, paracetamol
41.45 €
100mL
Estimated Shipment: 18-03-2025
to see stock.
Add to basket
 
10012831
View Documents
Macherey-Nagel™
814919
Aniline phthalate spray reagent 2-Propanol–Ethanol (1:1) Detection of reducing sugars, oxohalic acids
41.45 €
100mL
Please to purchase this item. Need a web account? Register with us today!
12450117
View Documents
Macherey-Nagel™
814922
4-(Dimethylamino)-benzaldehyde spray reagent 2-Propanol Detection of terpenes, sugars, steroids
41.45 €
1 set
Estimated Shipment: 21-03-2025
to see stock.
Add to basket
 
10276371
View Documents
Macherey-Nagel™
814403
Iron(III) Chloride spray reagent Water Detection of acetylsalicylic acid, paracetamol
41.45 €
100mL
Estimated Shipment: 18-03-2025
to see stock.
Add to basket
 
10736132
View Documents
Macherey-Nagel™
814401
Caffeine detection spray reagent Water-Acetone Caffeine-Detection
41.45 €
100mL
Please to purchase this item. Need a web account? Register with us today!
10206521
View Documents
Macherey-Nagel™
814923
Rhodamin B spray reagent Ethanol Detection of lipids
41.45 €
100mL
Please to purchase this item. Need a web account? Register with us today!
10246231
View Documents
Macherey-Nagel™
814206
Rubeanic acid spray reagent Ethanol Detection of heavy metal cations
41.45 €
100mL
Please to purchase this item. Need a web account? Register with us today!
12440117
View Documents
Macherey-Nagel™
814921
2'7'-Dichlorofluorescein spray reagent 2-Propanol Detection of lipids (saturated, unsaturated)
41.45 €
1 set
Please to purchase this item. Need a web account? Register with us today!
Description

Description

  • Broad selection of premium reagents available for detection of a wide range of substances
  • Can be used for spraying or dipping of TLC plates
  • Packaged in 100mL screw-top amber bottles
  • Use with chemically-resistant sprayers, including Macherey-Nagel glass laboratory sprayer with rubber bulb (mfr. no. 814101, sold separately)
Specifications

Specifications

7705-08-0
TLC plates
Spray Visualization Reagent
Solution
Spraying or Dipping
Amber bottle with screw cap
100 mL
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Ninhydrin spray reagent 100 ml

Signal Word
  • Danger

Hazard Category
  • Flammable liquid Category 2

Hazard Statement
  • H225-Highly flammable liquid and vapour.

Supplemental information
  • MIXTURE LIST-Contains : ethanol, ninhydrin

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.