missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Macherey-Nagel™ TLC Spray Visualization Reagents
Used for postchromatographic detection reactions in thin-layer chromatography. Macherey-Nagel™ TLC Spray Visualization Reagents are a selection of frequently used spray reagents that are suited for spraying or dipping TLC plates. Glass laboratory sprayer (mfr. no. 814101) sold separately.
Brand: Macherey-Nagel™ 814403
This item is not returnable.
View return policy
Description
- Broad selection of premium reagents available for detection of a wide range of substances
- Can be used for spraying or dipping of TLC plates
- Packaged in 100mL screw-top amber bottles
- Use with chemically-resistant sprayers, including Macherey-Nagel glass laboratory sprayer with rubber bulb (mfr. no. 814101, sold separately)
Specifications
7705-08-0 | |
Spraying or Dipping | |
1 to <2% Iron(III) Chloride | |
Detection of acetylsalicylic acid, paracetamol | |
Water | |
Odorless | |
4-5 | |
100 mL |
Slightly yellow | |
Iron(III) Chloride spray reagent | |
Iron(III) Chloride in Water | |
TLC plates | |
FeCl3 | |
Amber bottle with screw cap | |
Spray Visualization Reagent | |
Solution |