missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Kautex™ 250 mL Safety Wash Bottles Series 303 LDPE
Designed to give an accurate fine controlled stream of liquid. Kautex™ 250mL Safety Wash Bottles provide excellent chemical resistance for laboratory safety.
Brand: Kautex™ 2000770019
This item is not returnable.
View return policy
Description
- Imprinted with hazard symbols and substances of content in German, English and French language
- Writing space
- Abrasion-resistant printing
- Closure color matches color of ink for easy identification of media
- Leak-proof
- Completely emptied by sliding delivery tube, even when inverted.
Specifications
250 mL | |
30.8 mm | |
10 | |
Blue | |
Round | |
Wash Bottle | |
64 mm |
Natural | |
LDPE | |
150 Pk. | |
Wide | |
120 mm | |
Distilled Water | |
Screw Cap with Delivery Nozzle |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu