missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Kautex™ 250 mL Safety Wash Bottles Series 303 LDPE
Designed to give an accurate fine controlled stream of liquid. Kautex™ 250mL Safety Wash Bottles provide excellent chemical resistance for laboratory safety.
49.45 € - 753.95 €
Specifications
Capacity (Metric) | 250 mL |
---|---|
Color | Natural |
Diameter (Metric) Inner Neck | 30.8 mm |
Material | LDPE |
No. per Pack | 10 |
Product Code | Brand | Chemical Label | Closure Color | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Chemical Label | Closure Color | Price | Quantity & Availability | ||||
15758877
|
Kautex™
2000770020 |
Acetone | Red | N/A | |||||
15718877
|
Kautex™
2000770016 |
Ethanol | Green |
50.26 €
Pack of 10 |
Estimated Shipment: 20-01-2025
Log in to see stock. |
||||
15728877
|
Kautex™
2000770017 |
Isopropanol | Yellow | N/A | |||||
17858107
|
Kautex™
2000770018 |
Methanol | Orange | N/A | |||||
15738877
|
Kautex™
2000770018 |
Methanol | Orange |
49.45 €
Pack of 10 |
Estimated Shipment: 20-01-2025
Log in to see stock. |
||||
15748877
|
Kautex™
2000770019 |
Distilled Water | Blue |
50.26 €
Pack of 10 |
Estimated Shipment: 20-01-2025
Log in to see stock. |
||||
Description
- Imprinted with hazard symbols and substances of content in German, English and French language
- Writing space
- Abrasion-resistant printing
- Closure color matches color of ink for easy identification of media
- Leak-proof
- Completely emptied by sliding delivery tube, even when inverted.
Specifications
250 mL | |
30.8 mm | |
10 | |
Wide | |
120 mm | |
64 mm |
Natural | |
LDPE | |
150 Pk. | |
Round | |
Wash Bottle | |
Screw Cap with Delivery Nozzle |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title