Learn More
BRAND™ LDPE Pasteur Pipettes
LDPE Pasteur Pipettes are ideal for aliquots with very good reproducibility of the number of drops per milliliter.
50.85 € - 133.00 €
Specifications
Material LDPE
Quantity 500 Pack
Top Style Bulb Shaped
Type Pasteur Pipet
View More Specs
Products 6
Product Code Brand Length (Metric) Volume (Metric) Diameter (Metric) Outer Drops/mL Price Quantity & Availability  
Product Code Brand Length (Metric) Volume (Metric) Diameter (Metric) Outer Drops/mL Price Quantity & Availability  
15443594
View Documents
BRAND™
747775
144 mm 1 mL 1 mm 50 mL
133.00 €
Pack of 500
Estimated Shipment: 13-12-2024
to see stock.
Add to basket
10646341
View Documents
BRAND™
747765
152 mm 2 mL 3.3 mm 22 to 26
70.50 €
Pack of 500
In Stock
to see stock.
Edge
Add to basket
10562291
View Documents
BRAND™
747760
152 mm 3.5 mL 3.2 mm 21 to 28
50.85 €
Pack of 500
In Stock
to see stock.
Edge
Add to basket
10222661
View Documents
BRAND™
747755
150 mm 1 mL 3.4 mm 25 to 30
79.75 €
Pack of 500
In Stock
to see stock.
Edge
Add to basket
10100531
View Documents
BRAND™
747750
152 mm 3 mL 2.8 mm 25 to 27
101.00 €
Pack of 500
In Stock
to see stock.
Edge
Add to basket
10721963
View Documents
BRAND™
747770
150 mm 4 mL 3 mm 60 to 75
130.00 €
Pack of 500
In Stock
to see stock.
Edge
Add to basket
Description
Description
  • Manufactured from LDPE.
  • Very good reproducibility of the number of drops per milliliter.
  • Ideal for aliquots.
  • Pasteur pipettes can be filled and deep-frozen, or changed into a closed vessel by heat-sealing the tip.
  • The integrated pipetting bulb depresses easily, minimizing fatigue from frequent pipetting.
  • Resistant to gas or gamma radiation sterilization.
Specifications
Specifications
LDPE
500 Pack
Bulb Shaped
Pasteur Pipet