missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lead(Ii) Carbonate Basic, Honeywell Fluka™
Puriss., white lead,≥77% Pb basis
129.00 € - 5439.17 €
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15691050
|
Honeywell Research Chemicals
11513-1KG |
1 kg |
129.00 €
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15611060
|
Honeywell Research Chemicals
|
6 x 1 kg |
644.97 €
Pack of 6 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15601060
|
Honeywell Research Chemicals
11513-50KG |
50 kg |
5439.17 €
50kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Chemical Identifiers
1319-46-6 | |
MFCD00078155 |
Specifications
1319-46-6 | |
MFCD00078155 | |
775.63g/mol | |
puriss. |
(PbCO3)2·Pb(OH)2 | |
3077 | |
≥77% (Pb basis) | |
Lead(Ii) Carbonate Basic |
Safety and Handling
P201-P260-P280-P301 + P312 + P330-P308 + P313
H302 + H332-H360Df-H373-H410
missing translation for 'einecsNumber' : 215-290-6
missing translation for 'rtecsNumber' : OF9275000
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- Lead(II) carbonate basic
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 1
- ACUTE AQUATIC Acute 1
- Reproductive toxicity Category 1A
- Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
- H302-Harmful if swallowed.
- H332-Harmful if inhaled.
- H360Df-May damage the unborn child. Suspected of damaging fertility.
- H373-May cause damage to organs.
- H400-Very toxic to aquatic life.
- H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P284-¢In case of inadequate ventilation! wear respiratory protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
- MIXTURE LIST-Contains : Lead(II) carbonate basic