Learn More
Lead(II) carbonate, White powder, Puratronic™, 99.999% (Metals basis), Thermo Scientific Chemicals
47.50 € - 320.00 €
Chemical Identifiers
CAS | 598-63-0 |
---|---|
Molecular Formula | CO3Pb |
Molecular Weight (g/mol) | 267.20 |
MDL Number | MFCD00064631 |
InChI Key | MFEVGQHCNVXMER-UHFFFAOYSA-L |
Synonym | lead carbonate, lead ii carbonate, cerussete, cerussite, lead 2+ carbonate, plumbous carbonate, dibasic lead carbonate, unii-43m0p24l2b, carbonic acid, lead 2+ salt 1:1, lead carbonate pbco3 |
PubChem CID | 11727 |
SMILES | [Pb++].[O-]C([O-])=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11306797
|
Thermo Scientific Alfa Aesar
010721.06 |
5 g | |||||||
11316797
|
Thermo Scientific Alfa Aesar
010721.14 |
25 g |
320.00 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
598-63-0 | |
267.20 | |
MFEVGQHCNVXMER-UHFFFAOYSA-L | |
11727 |
CO3Pb | |
MFCD00064631 | |
lead carbonate, lead ii carbonate, cerussete, cerussite, lead 2+ carbonate, plumbous carbonate, dibasic lead carbonate, unii-43m0p24l2b, carbonic acid, lead 2+ salt 1:1, lead carbonate pbco3 | |
[Pb++].[O-]C([O-])=O |
Specifications
598-63-0 | |
Powder | |
CO3Pb | |
lead carbonate, lead ii carbonate, cerussete, cerussite, lead 2+ carbonate, plumbous carbonate, dibasic lead carbonate, unii-43m0p24l2b, carbonic acid, lead 2+ salt 1:1, lead carbonate pbco3 | |
[Pb++].[O-]C([O-])=O | |
267.20 | |
267.2 | |
6.6g/cm3 at 20°C |
White | |
99.999% (Metals basis) | |
MFCD00064631 | |
MFEVGQHCNVXMER-UHFFFAOYSA-L | |
λ²-lead(2+) carbonate | |
11727 | |
Odorless | |
Lead(II) carbonate |
Safety and Handling
Avoid exposure - obtain special instructions before use. In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible). This material and/or its container must be disposed of as hazardous waste. Avoid release to the environment. Refer to special instructions/Safety data sheets.
H360-H373-H400-H410-H302-H332: May damage fertility or the unborn child. May cause damage to organs through prolonged or repeated exposure. Very toxic to aquatic life. Very toxic to aquatic life with long lasting effects. Harmful if swallowed. Harmful if inhaled.
missing translation for 'dotInformation' : DOT Class: 6.1, Packaging Group: III
missing translation for 'einecsNumber' : 209-943-4
- Lead(II) carbonate
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- Reproductive toxicity Category 1A
- Specific target organ toxicity after repeated exposure Category 2
- ACUTE AQUATIC Acute 1
- LONG-TERM AQUATIC HAZARD Chronic 1
Hazard Statement
- H302-Harmful if swallowed.
- H332-Harmful if inhaled.
- H360Df-May damage the unborn child. Suspected of damaging fertility.
- H373-May cause damage to organs.
- H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P405-Store locked up.
- P501-Dispose of contents/container to .
RUO – Research Use Only