Learn More
Lead(II) iodide, 99%, Thermo Scientific Chemicals
76.40 € - 249.00 €
Chemical Identifiers
CAS | 10101-63-0 |
---|---|
Molecular Formula | I2Pb |
Molecular Weight (g/mol) | 461.00 |
MDL Number | MFCD00011163 |
InChI Key | RQQRAHKHDFPBMC-UHFFFAOYSA-L |
Synonym | lead ii iodide, lead diiodide, plumbous iodide, lead iodide pbi2, pbi2, plumbum jodatum, blei ii-iodid, hsdb 636, diiodo-, e2-plumbane |
PubChem CID | 24931 |
IUPAC Name | λ²-lead(2+) diiodide |
SMILES | [I-].[I-].[Pb++] |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10123740
|
Thermo Scientific Acros
198860500 |
50 g | Glass bottle |
76.40 €
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10655272
|
Thermo Scientific Acros
198862500 |
250 g | Glass bottle |
249.00 €
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
10101-63-0 | |
461.00 | |
RQQRAHKHDFPBMC-UHFFFAOYSA-L | |
24931 | |
[I-].[I-].[Pb++] |
I2Pb | |
MFCD00011163 | |
lead ii iodide, lead diiodide, plumbous iodide, lead iodide pbi2, pbi2, plumbum jodatum, blei ii-iodid, hsdb 636, diiodo-, e2-plumbane | |
λ²-lead(2+) diiodide |
Specifications
402°C | |
Yellow to Orange | |
98.5% min. (Argentometry) | |
PbI2 | |
15, 5466 | |
RQQRAHKHDFPBMC-UHFFFAOYSA-L | |
λ²-lead(2+) diiodide | |
24931 | |
99% | |
6.16 | |
Lead(II) iodide, 99% |
10101-63-0 | |
Powder | |
I2Pb | |
MFCD00011163 | |
lead ii iodide, lead diiodide, plumbous iodide, lead iodide pbi2, pbi2, plumbum jodatum, blei ii-iodid, hsdb 636, diiodo-, e2-plumbane | |
[I-].[I-].[Pb++] | |
461.00 | |
461 | |
0.1% max. | |
6.1600g/mL |
Safety and Handling
GHS H Statement
May cause damage to organs through prolonged or repeated exposure.
Very toxic to aquatic life with long lasting effects.
Harmful if inhaled.
May damage the unborn child.
Suspected of damaging fertility.
GHS P Statement
Use personal protective equipment as required.
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for b
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 233-256-9
missing translation for 'tsca' : TSCA
- Lead(II) iodide
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 1
- ACUTE AQUATIC Acute 1
- Reproductive toxicity Category 1A
- Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
- H400-Very toxic to aquatic life.
- H410-Very toxic to aquatic life with long lasting effects.
- H360Df-May damage the unborn child. Suspected of damaging fertility.
- H373-May cause damage to organs.
- H332-Harmful if inhaled.
- H302-Harmful if swallowed.
Precautionary Statement
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
RUO – Research Use Only