Learn More
Lead(II) nitrate, ACS, 99.0% min, Thermo Scientific Chemicals
For Use As An Oxidizer in Dye Industry, in Manufacturing of Mother-of-Pearl, As Mordant in Printing Textile
54.27 € - 149.75 €
Chemical Identifiers
CAS | 10099-74-8 |
---|---|
Molecular Formula | N2O6Pb |
Molecular Weight (g/mol) | 331.20 |
MDL Number | MFCD00011153 |
InChI Key | RLJMLMKIBZAXJO-UHFFFAOYSA-N |
Synonym | lead dinitrate, lead nitrate, lead ii nitrate, plumbous nitrate, lead 2+ nitrate, lead nitrate pb no3 2, nitrate de plomb french, lead ii nitrate 1:2, nitric acid, lead 2+ salt |
PubChem CID | 24924 |
ChEBI | CHEBI:37187 |
SMILES | [Pb++].[O-][N+]([O-])=O.[O-][N+]([O-])=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11307817
|
Thermo Scientific Alfa Aesar
014243.36 |
500 g |
54.27 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11397807
|
Thermo Scientific Alfa Aesar
014243.A3 |
2 kg |
149.75 €
2kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
As an oxidizer in dye industry, in manufacturing of mother-of-pearl, as mordant in printing textile, as sensitizer in photographyEmployed in industrial applications such as heat stabilization in nylon and polyesters, coatings of photothermographic paper, gold cyanidation and pyrotechnics. Finds
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
10099-74-8 | |
331.20 | |
RLJMLMKIBZAXJO-UHFFFAOYSA-N | |
24924 | |
[Pb++].[O-][N+]([O-])=O.[O-][N+]([O-])=O |
N2O6Pb | |
MFCD00011153 | |
lead dinitrate, lead nitrate, lead ii nitrate, plumbous nitrate, lead 2+ nitrate, lead nitrate pb no3 2, nitrate de plomb french, lead ii nitrate 1:2, nitric acid, lead 2+ salt | |
CHEBI:37187 |
Specifications
470°C (decomposition) | |
White | |
99.0% min. | |
MFCD00011153 | |
Freely soluble in water. Soluble in absolute alcohol,methanol. Insoluble in concentrated HNO3 | |
[Pb++].[O-][N+]([O-])=O.[O-][N+]([O-])=O | |
331.20 | |
CHEBI:37187 | |
ACS | |
Hygroscopic | |
approx.3.0-4.0 (20% aqueous solution) | |
Lead(II) nitrate, For ACS analysis |
10099-74-8 | |
Crystals | |
N2O6Pb | |
lead dinitrate, lead nitrate, lead ii nitrate, plumbous nitrate, lead 2+ nitrate, lead nitrate pb no3 2, nitrate de plomb french, lead ii nitrate 1:2, nitric acid, lead 2+ salt | |
RLJMLMKIBZAXJO-UHFFFAOYSA-N | |
lead(2+);dinitrate | |
24924 | |
331.2 | |
Odorless | |
White crystalline | |
4.53 g/cm3 at 20°C |
Safety and Handling
P201-P202-P210-P220-P221-P260-P264b-P270-P271-P281-P301+P312-P304+P340-P305+P351+P338-P308+P313-P310-P330-P370+P378q-P501c
H272-H302+H332-H318-H336-H350-H360Df-H373
missing translation for 'dotInformation' : DOT Class: 5.1, Packaging Group: II
missing translation for 'einecsNumber' : 233-245-9
missing translation for 'rtecsNumber' : OG2100000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
- Lead(II) nitrate
Signal Word
- Danger
Hazard Category
- Oxidising solid Category 2
- Acute toxicity Category 4
- Reproductive toxicity Category 1A
- Specific target organ toxicity after repeated exposure Category 2
- ACUTE AQUATIC Acute 1
- LONG-TERM AQUATIC HAZARD Chronic 1
Hazard Statement
- H272-May intensify fire; oxidiser.
- H302-Harmful if swallowed.
- H332-Harmful if inhaled.
- H360Df-May damage the unborn child. Suspected of damaging fertility.
- H373-May cause damage to organs.
- H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P201-Obtain special instructions before use.
- P220-Keep/Store away from clothing/combustible materials.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P405-Store locked up.
- P501-Dispose of contents/container to .
RUO – Research Use Only