missing translation for 'onlineSavingsMsg'
Learn More

Lead(II) oxide, Puratronic™, 99.999% (metals basis), Thermo Scientific Chemicals

87.33 € - 842.19 €

Chemical Identifiers

CAS 1317-36-8
Molecular Formula OPb
Molecular Weight (g/mol) 223.20
MDL Number MFCD00011164
InChI Key YEXPOXQUZXUXJW-UHFFFAOYSA-N
Synonym lead monoxide, lead ii oxide, lead oxide pbo, massicot, massicotite, lead monooxide, lead protoxide, plumbous oxide, litharge pure, lead oxide yellow
PubChem CID 14827
IUPAC Name oxolead
SMILES O=[Pb]
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11355047
View Documents
Thermo Scientific Alfa Aesar
040753.14
25 g
87.33 €
25g
Please to purchase this item. Need a web account? Register with us today!
11365047
View Documents
Thermo Scientific Alfa Aesar
040753.22
100 g
246.23 €
100g
Please to purchase this item. Need a web account? Register with us today!
11348678
View Documents
Thermo Scientific Alfa Aesar
040753.36
500 g
842.19 €
500g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Used in producing iridescent colors on brass and bronze, coloring sulfur-containing substances and used as pigment for rubber. It is also used in oil refining, varnishes, paints, enamels and in assays for precious metal ores.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

1317-36-8
223.20
YEXPOXQUZXUXJW-UHFFFAOYSA-N
14827
O=[Pb]
OPb
MFCD00011164
lead monoxide, lead ii oxide, lead oxide pbo, massicot, massicotite, lead monooxide, lead protoxide, plumbous oxide, litharge pure, lead oxide yellow
oxolead

Specifications

886°C
Yellow
OPb
UN3077
lead monoxide, lead ii oxide, lead oxide pbo, massicot, massicotite, lead monooxide, lead protoxide, plumbous oxide, litharge pure, lead oxide yellow
YEXPOXQUZXUXJW-UHFFFAOYSA-N
oxolead
14827
99.999%
Odorless
(metals basis)
1317-36-8
Powder
MFCD00011164
14,5413
Insoluble in water,soluble in concentrated alkalies,hydrochloric acid and ammonium chloride.
O=[Pb]
223.20
223.19
Puratronic™
9.53 g/mL
Lead(II) oxide
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H360-H373-H302-H332
May damage fertility or the unborn child.
May cause damage to organs through prolonged or repeated exposure.
Harmful if swallowed.
Harmful if inhaled.

P201-P202-P260-P264b-P270-P271-P281-P301+P312-P304+P340-P308+P313-P330-P501c

H302+H332-H350-H360Df-H372

missing translation for 'dotInformation' : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.

missing translation for 'einecsNumber' : 215-267-0

missing translation for 'rtecsNumber' : OG1750000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

Product Identifier
  • Lead(II) oxide

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.