missing translation for 'onlineSavingsMsg'
Learn More

Bel-Art™ SP Scienceware™ Weighted Beaker/Flask Holders

Weighted holder grips beakers or flasks firmly with four flared stainless steel prongs.

Brand:  Bel-Art™ SP Scienceware™ F18309-0100

149.98 EUR valid until 2025-04-30
Use promo code "24138" to get your promotional price.


 View more versions of this product

Product Code. 12633056

  • 159.83 € / Each
Estimated Shipment: 18-06-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Vinyl-coated lead ring weight secured beneath holder
  • Keeps container immersed in water
  • Provides a low center of gravity
  • Allows water to circulate underneath the beaker or flask
  • Can be used in boiling liquid up to 135°C (275°F) for 1 hour
Specifications

Specifications

Weighted Beaker
Lead
1 Each
135°C
208 g
Red
100mL Beakers and Flasks
6 mm
6 cm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

One moment while we fetch your results.
Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Bel-Art™ SP Scienceware™ Weighted Beaker/Flask Holders > Outer Diameter: 6mm

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.