Learn More
Fisherbrand™ Leaf Calibration Weight Set, Class M1 with certificate
Use with weighing instruments of medium accuracy class III. Fisherbrand™ Class M1 Stainless Steel Calibration Weights are ideal for the verification or calibration of class M2 weights.
Brand:  Fisherbrand™ FB67830
Product Code. 15595109
  • 184.00 € / 1 unit
Estimated Shipment: 11-06-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Class M1 calibration weight
  • Delivered with a calibration certificate guaranteeing traceability
  • In compliance with OIML R111
  • Supplied individually or as a set, in a plastic case or a wooden box
Specifications
Specifications
M1
1 to 500 mg
1 mg to 500 mg
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Fisherbrand™ Leaf Calibration Weight Set, Class M1 with certificate > 1mg to 500mg