Learn More
Lecithin, MedChemExpress
MedChemExpress Lecithin is regarded as a safe, conventional phospholipid source. Phospholipids are reported to alter the fatty acid composition and microstructure of the membranes in animal cells.
Brand:  Medchem Express HY-B2235
Encompass_Preferred
 View more versions of this product
Product Code. 16468876
  • 70.20 € / 100mg
Explore more special offers
Description
Description
Lecithin is regarded as a safe, conventional phospholipid source. Phospholipids are reported to alter the fatty acid composition and microstructure of the membranes in animal cells.
Specifications
Specifications
Lecithin
Light Yellow
C42H80NO8P
DMSO : 5 mg/mL (6.60 mM; Need ultrasonic) ∣H2O : 3.33 mg/mL (4.39 mM; Need ultrasonic)
758.06
98.0%
Solid
8002-43-5
Research
100 mg
O=P(OC[C@@H](COC(CCCCCCCCCCCCCCC)=O)OC(CCCCCCC/C=C\C/C=C\CCCCC)=O)(OCC[N+](C)(C)C)[O-]
758.06
Research