Learn More
Cole-Parmer™ Leica Microsystems™ Platinum Calibrated Loops
Precisely designed calibrated loop suitable for milk samples. Cole-Parmer™ Leica Microsystems™ Platinum Calibrated Loops are fused to form a perfect circle for accurate delivery.
Brand:  Cole-Parmer™ 01850-02
Product Code. 11785868
  • 280.00 € / Each
Estimated Shipment: 27-05-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Ideal for milk cultures.
  • Calibrated to American Public Health Association (APHA) standards and made of B&S platinum wire with 5% rhodium
  • Insulated loop and needle holder available separately
Specifications
Specifications
10 μL
25 mm
Transferring bacterial cultures
Yes
4 mm
19-ga B&S Platinum Wire
19
Transferring bacterial cultures
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Cole-Parmer™ Leica Microsystems™ Platinum Calibrated Loops > Volume: 10uL