Learn More
Fisherbrand™ Lid for Desiccator with Glass Knob
For use with desiccators with glass knob
Brand:  Fisherbrand™ 632415280301
 View more versions of this product
Product Code. 15514645
  • 169.00 € / Each
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
  • For use with desiccators with glass knob
Certifications
ISO 13 130, DIN 12 491
Specifications
Specifications
380 mm
Type: 300 mm
150 mm
Lid
Desiccator with glass knob
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Fisherbrand™ Lid for Desiccator with Glass Knob > Type: 300mm; Diameter: 380mm; Height: 150mm