missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Lid for Desiccator with Glass Knob
For use with desiccators with glass knob
77.45 € - 169.00 €
Product Code | Brand | Diameter (Metric) Outer | Height (Metric) | Description | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) Outer | Height (Metric) | Description | Price | Quantity & Availability | ||||
15564635
|
Fisherbrand™
632415280251 |
320 mm | 125 mm | Type: 250 mm |
118.00 €
Each |
Estimated Shipment: 15-05-2025
Log in to see stock. |
||||
15564625
|
Fisherbrand™
632415280151 |
215 mm | 103 mm | Type: 150 mm |
106.00 €
Each |
Estimated Shipment: 15-05-2025
Log in to see stock. |
||||
15514645
|
Fisherbrand™
632415280301 |
380 mm | 150 mm | Type: 300 mm |
169.00 €
Each |
In Stock
Log in to see stock. |
||||
15514635
|
Fisherbrand™
632415280201 |
270 mm | 108 mm | Type: 200 mm |
96.30 €
Each |
Estimated Shipment: 02-05-2025
Log in to see stock. |
||||
15514625
|
Fisherbrand™
632415280101 |
153 mm | 78 mm | Type: 100 mm |
77.45 €
Each |
Estimated Shipment: 15-05-2025
Log in to see stock. |
||||
Description
- For use with desiccators with glass knob
Certifications
ISO 13 130, DIN 12 491
Specifications
Lid | |
Desiccator with glass knob |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Fisherbrand™ Lid for Desiccator with Glass Knob