missing translation for 'onlineSavingsMsg'
Learn More

Labconco™ Lids for FreeZone™ Freeze Dryers

Replacement lids for FreeZone™ Freeze Dryer collector chambers.

Brand:  Labconco™ 7208200

 View more versions of this product

Product Code. 16328816

  • 216.00 € / Each
Estimated Shipment: 24-06-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Stainless Steel, 14 in.

  • Stainless steel lid has 14.0 in. diameter with 3 in. diameter hole
  • Optional collector lid for FreeZone™ 4.5L, 6L, 8L, 12L and 18L Freeze Dryers
  • For use with solvents that attack arcylic, like acetonitrile
  • Includes neoprene gasket
Specifications

Specifications

Chamber Lid
FreeZone™ 6L, 12L, and 18L Console Freeze Dryers
14.0 in.
3.18 cm
36.2 cm
2.5 lb.
Acrylic
Neoprene gasket
1.25 in.
14.25 in.
5 lb.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Labconco™ Lids for FreeZone™ Freeze Dryers > Acrylic; 14.0 in. diameter

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.