Learn More
Lenz Laborglasintrumente™ Liebig Condenser with GL Threads
Liebig condenser for condensing vapours
Brand:  Lenz Laborglasintrumente™ 05210314
Product Code. 10502503
  • 48.00 € / Each
Estimated Shipment: 21-05-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Liebig condensers acc. to DIN 12576 and in addition to DIN
  • With NS socket and NS cone
  • Water connection: GL 14 threads (with two threaded plastic olives)
  • Made of DURAN™ tubing
Specifications
Specifications
DIN 12576
250 mm
NS 29/32
Cooling Surface - 130 cm²
1
DURAN™ Tubing
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Lenz Laborglasintrumente™ Liebig Condenser with GL Threads > Standard Taper Joint No.: NS 29/32