Learn More
3M™ Powerflow™ Turbo Unit PFT-01
3M™ Powerflow™ Turbo Unit is designed for use with 3M™ Powerflow™ Powered Air Respirator 7900PF.
Brand:  3M™ 7000104113
Product Code. 15739897
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
  • Replacement powered air respirator turbo unit
  • Compact face-mounted turbo unit with DIN knuckle threads
  • Genuine 3M™ Accessory
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Turbo Unit
3M™ Powerflow™ Powered Air Respirator 7900PF
Product Suggestions
Product Suggestions
Videos
Videos
  • 3M - Powered and supplied air products
    Video Player is loading.
    Current Time 0:00
    Duration 0:00
    Loaded: 0%
    Stream Type LIVE
    Remaining Time 0:00
    1x
    • Chapters
    • descriptions off, selected
    • captions off, selected
      3M Powered and supplied air products
    SDS
    Documents
    Documents
    Certificates
    Special Offers
    Special Offers
    Product Content Correction
    Product Title
    3M™ Powerflow™ Turbo Unit PFT-01 > Product Type: Turbo Unit