missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Purolite™ Lifetech™ ECR Resins for Enzyme Immobilization, 150 to 300 μm
Lifetech™ ECR resins are robust polymeric resins designed for research and industrial immobilization of enzymes. Available for covalent, ionic, adsorption and affinity immobilization.
Brand: Purolite™ LS01211-94
This item is not returnable.
View return policy
Description
- Enzyme immobilization by adsorption, covalent, IEX and affinity
- High mechanical stability
- High porosity
- Easily dried after immobilization
- Suitable for batch and column configuration
- Optimal for use in organic solvent/water free systems
- Compliant with ResAP(2004) 3
- Halal, Kosher and TSE/BSE/GMO free
Specifications
5 to 9 | |
2°C to 8°C | |
50 to 65% | |
Wet | |
1 kg | |
White to Off-White Spherical Beads |
300 to 600 Å | |
150 to 300 μm | |
Epoxy | |
Lifetech™ | |
Methacrylate (ECR Resin) |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction