missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Purolite™ Lifetech™ ECR Resins for Enzyme Immobilization, 150 to 300 μm
Lifetech™ ECR resins are robust polymeric resins designed for research and industrial immobilization of enzymes. Available for covalent, ionic, adsorption and affinity immobilization.
241.00 € - 825.00 €
Specifications
Particle Size | 150 to 300 μm |
---|---|
Form | Wet |
Product Line | Lifetech™ |
Type | Methacrylate (ECR Resin) |
Format | White to Off-White Spherical Beads |
Product Code | Brand | Quantity | Pore Size | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Pore Size | Price | Quantity & Availability | ||||
17009333
|
Purolite™
LS01231-192 |
250 g | 600 to 1200 Å | N/A | |||||
17009363
|
Purolite™
LS01361-192 |
250 g | 1200 to 1800 Å | N/A | |||||
17019333
|
Purolite™
LS01231-94 |
1 kg | 600 to 1200 Å | N/A | |||||
17019353
|
Purolite™
LS01351-192 |
250 g | 600 to 1200 Å | N/A | |||||
17019363
|
Purolite™
LS01361-94 |
1 kg | 1200 to 1800 Å | N/A | |||||
17029353
|
Purolite™
LS01351-94 |
1 kg | 600 to 1200 Å | N/A | |||||
17029373
|
Purolite™
LS01381-192 |
250 g | 600 to 1200 Å | N/A | |||||
17039373
|
Purolite™
LS01381-94 |
1 kg | 600 to 1200 Å | N/A | |||||
17049323
|
Purolite™
LS01211-192 |
250 g | 300 to 600 Å | N/A | |||||
17059323
|
Purolite™
LS01211-94 |
1 kg | 300 to 600 Å | N/A | |||||
17059343
|
Purolite™
LS01311-192 |
250 g | 300 to 600 Å | N/A | |||||
17069333
|
Purolite™
LS01241-192 |
250 g | 1200 to 1800 Å | N/A | |||||
17069343
|
Purolite™
LS01311-94 |
1 kg | 300 to 600 Å | N/A | |||||
17069363
|
Purolite™
LS01371-192 |
250 g | 300 to 600 Å | N/A | |||||
17079333
|
Purolite™
LS01241-94 |
1 kg | 1200 to 1800 Å | N/A | |||||
17079363
|
Purolite™
LS01371-94 |
1 kg | 300 to 600 Å | N/A | |||||
17089373
|
Purolite™
LS01391-192 |
250 g | 1200 to 1800 Å | N/A | |||||
17099373
|
Purolite™
LS01391-94 |
1 kg | 1200 to 1800 Å | N/A | |||||
Description
- Enzyme immobilization by adsorption, covalent, IEX and affinity
- High mechanical stability
- High porosity
- Easily dried after immobilization
- Suitable for batch and column configuration
- Optimal for use in organic solvent/water free systems
- Compliant with ResAP(2004) 3
- Halal, Kosher and TSE/BSE/GMO free
Specifications
150 to 300 μm | |
Lifetech™ | |
White to Off-White Spherical Beads |
Wet | |
Methacrylate (ECR Resin) |
Safety and Handling
missing translation for 'shelfLife' : Use within 6 months from date of manufacture
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title