Learn More
Fisherbrand™ Mini Light Box
Store it in a drawer to save work space. Fisherbrand™ Mini Light Box is perfect for quick screening of well plates, Petri dishes, gels, slides and transparencies. The battery operated unit keeps illumination within arm's reach without cluttering a crowded benchtop.
Brand:  Fisherbrand™ HS15093
Product Code. 12852869
Description
Description
  • Perfect for space-challenged labs or classrooms
  • Can be stored in a drawer or with limited footprint on the benchtop
  • 10 x 127mm viewing area is ideal for 96-well plates, Petri dishes, electrophoresis gels, slides and transparencies
  • Powered by four AA batteries (included) for a portable light box
Specifications
Specifications
AAA
44 mm
135 mm
96-Well Plates, Petri Dishes, Electrophoresis Gels, Slides and Transparencies
161 mm
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
View accessory products
Special Offers
Special Offers
Product Content Correction
Product Title
Fisherbrand™ Mini Light Box > Dimensions: 161 x 135 x 44mm