Learn More
Cole-Parmer™ Lightnin Heavy-Duty Gear Drive Mixer
Lightnin Heavy-Duty Gear Drive Mixers are ideal for mixing your high-viscosity materials or large batches of water-like fluids. All models maintain speeds from 20 to 350 rpm (20:1 ratio) and generate enough torque to mix highly viscous fluids.
Brand:  Cole-Parmer™ 50310-46
 View more versions of this product
Product Code. 15467453
  • 4535.00 € / Each
Estimated Shipment: 26-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Includes:
    • 316 SS shaft, 316 SS A-310 impeller, and drive quill with set screw.
    • Models 50310-68, -70 and -72 also include bung adapter and a folding impeller.
  • More About This Item:
    • These variable-speed, gear-drive mixers are ideal for mixing your high-viscosity materials or large batches of water-like fluids. All models maintain speeds from 20 to 350 rpm (20:1 ratio) and generate enough torque to mix highly viscous fluids.
    • Gear-drive mixers mount with 20°> offset tank clamp (included). Available as either an air-powered or electric mixer.
    • Select from three speed options; 20 to 350 rpm (20:1 ratio), 20 to 280 rpm (14:1 ratio), or fixed speed of 350 or 280 rpm. The 3⁄4-hp mixers generate plenty of torque for your high-viscosity applications.
Specifications
Specifications
20 to 350 rpm
19.05 mm
316 Stainless Steel
XPRF
0.75 in.
316 SS Shaft, 316 SS A-310 Impeller, and Drive Quill with Set Screw
EV5P25ET1F48
720 in. oz.