missing translation for 'onlineSavingsMsg'
Learn More
Lithium chloride, 99+%, ACS reagent, Thermo Scientific Chemicals Product Code.: 10328823

Lithium chloride, 99+%, ACS reagent, Thermo Scientific Chemicals

Product Code. 10328823
100 g, Plastic bottle
Click to view available options
Quantity:
5 g
100 g
500 g
2.5 kg
Packaging:
Glass Bottle
Plastic bottle
This item is not returnable. View return policy

Product Code. 10328823

Brand: Thermo Scientific Acros 413271000

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7447-41-8
Molecular Formula ClLi
Molecular Weight (g/mol) 42.39
MDL Number MFCD00011078
InChI Key KWGKDLIKAYFUFQ-UHFFFAOYSA-M
Synonym lithium chloride, lithium chloride licl, lithiumchloride, licl, chlorure de lithium, chlorku litu, chlorolithium, lithiumchlorid, chlorku litu polish, luthium chlorideShow MoreShow Less
PubChem CID 433294
ChEBI CHEBI:48607
SMILES [Li+].[Cl-]

Specifications

Boiling Point 1382.0°C
Melting Point 605.0°C
Color White
Physical Form Crystalline Powder and/or Crystals
Quantity 100 g
Assay Percent Range 99% min. (Argentometry)
Linear Formula LiCl
Fieser 01,609; 02,246; 04,298; 05,677; 12,277; 16,194
Merck Index 15, 5584
Solubility Information Solubility in water: 832g/L (20°C). Other solubilities: soluble in alcohols,ether,pyridine,,nitrobenzene and acetone
IUPAC Name lithium(1+) chloride
Formula Weight 42.38
Percent Purity ≥99%
Grade ACS Reagent
Loss on Drying 1.0% max. (1 g, 105°C, 2 hrs)
Packaging Plastic bottle
Specific Gravity 2.06
Density 2.0600g/mL
Chemical Name or Material Lithium chloride, 99%
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0451530 Certificate TypeCertificate of Analysis Date17/04/2024 Catalog Number413270025,413271000,413275000,413270050
Lot NumberA0460076 Certificate TypeCertificate of Analysis Date13/02/2024 Catalog Number413270025,413271000,413275000,413270050
Lot NumberA0454604 Certificate TypeCertificate of Analysis Date08/02/2024 Catalog Number413270025,413271000,413275000,413270050
Lot NumberA0459857 Certificate TypeCertificate of Analysis Date02/02/2024 Catalog Number413270025,413271000,413275000,413270050
Lot NumberA0459858 Certificate TypeCertificate of Analysis Date02/02/2024 Catalog Number413270025,413271000,413275000,413270050
compliance-icons
Product Identifier
  • Lithium chloride, anhydrous
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Lithium chloride, 99+%, ACS reagent, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.