Learn More
AccuGENE™ Molecular Biology Water, Lonza™
Ready-to-use for a wide range of molecular biology applications. AccuGENE™ Molecular Biology Water is prepared with 18 megOhm water, filtered using a 0.2 micron filter, and filled into sterile bottles.
49.00 €
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11548015
|
Lonza
BE51200 |
1 L | N/A | ||||||
Description
- Reliable - Manufactured according to strict quality control standards to ensure lot-to-lot consistency
- High Quality - No detectable DNase, RNase, or protease activities
- Efficient - Ready-made solutions eliminate experiment preparation time
- Flexible - Customized solutions are available to meet individual needs
For research or manufacturing use only.