Learn More
Lovibond™ ET 740 and ET 750 - Laboratory Floc / Jar Tester
Flocculation Testers (also known as Jar Testers) are designed for a range of applications - such as testing the efficiency of flocculation or precipitation agents
Brand:  Lovibond™ 2419160
 View more versions of this product
Product Code. 12661745
  • 2820.00 € / Each
Estimated Shipment: 17-12-2024
to see stock.
Explore more special offers
Add to basket
Includes: Power Supply with US, EU and UK adapter. Instruction Manual.
Requires : 1,000 mL beakers, Low form
Description
Description
Flocculation Testers (also known as Jar Testers) are designed for a range of applications - such as testing the efficiency of flocculation or precipitation agents
Continuously variable stirring speed (lab versions only.)
  • Digital display of stirring RPM.
  • Timer function.
  • Illuminated back panel.
  • Height adjustment option for the stirring blades during operation.
  • Date and Time.
Floc Testers are not supplied with beakers. Please purchase from a laboratory products supplier.
Specifications
Specifications
Laboratory Flocculation Tester
Power supply with US, EU and UK adapter, instruction manual
For a range of applications such as testing the efficiency of flocculation or precipitation agents
37 x 13.5 x 10.25 in. (93.9 x 34.2 x 26.03 cm)
93.9 cm
34.2 cm
26.03 cm
50/60 Hz
37.5 lb.
Digital
Timer
1 Revolution
37 in.
13.5 in.
10.25 in.
115/230 V 50/60 Hz
115/230 V
17 kg