missing translation for 'onlineSavingsMsg'
Learn More

Lovibond™ Model 711-A Nesslerizer System Nessler Tubes

Set of two clear glass Nessler tubes

Brand:  Lovibond™ 354200

Product Code. 10668342

  • 88.40 € / Pack of 2
Estimated Shipment: 08-08-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Water sample tubes
  • Path length: 250 mm (9.8 in.)
  • Plungers included
  • Use for water color comparison
  • Compatible with: Lovibond Nessslerizer System 711-A1 (FS Cat. No. 15-405-311)

Specifications

Specifications

Glass
250 mm
Water color comparison
Clear
711-A1 Orbeco-Hellige™/Lovibond™ nessslerizer system
Plungers
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Lovibond™ Model 711-A Nesslerizer System Nessler Tubes > Nessler Tubes; 250mm (9.8 in.); Set of two

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.