Learn More
Thermo Scientific™ Standard Fluid Transfer Assembly Y Set
Simplify process connectivity and enable greater efficiency for most upstream and downstream process operations.
Brand:  Thermo Scientific™ SH31096.01
 View more versions of this product
Product Code. 17880627
  • 486.00 € / Pack of 10
Explore more special offers
Description
Description
Standard Fluid Transfer Assembly products interconnect with Thermo Scientific standard Labtainer BPCs, Productainer BPCs, single-use bioreactors, mixers, and fermentors right on the shop floor to deliver a comprehensive and complete set of single-use products to meet the needs of your facility or process. Designed to aid many common shop floor processes such as sampling, transfer, and filling, a fit-for-purpose solution in the required tubing size and length can be easily selected from these designs.
  •  One set of stocked products reduces the number of customized stock keeping units (SKUs), simplifies the ordering process, and reduces product storage requirements
  • Multiple manufacturing sites ensure security in product supply
  • Produced under cGMP conditions in ISO 7 clean areas to ensure a high level of quality
  • Characterized using ISO, USP, EP, and ASTM test methods to validate performance
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Approved for storage at ambient conditions (15°C to 30°C).
3 x 12.7mm (1/2 in) ID x 3.2mm (1/8 in) wall, C-Flex 374, 2 x 0.51m (20 in) to 0.51m (20 in)
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Thermo Scientific™ Standard Fluid Transfer Assembly Y Set > 3 x 12.7mm (1/2 in) ID x 3.2mm (1/8 in) wall, C-Flex 374, 2 x 0.51m (20 in) to 0.51m (20 in)