missing translation for 'onlineSavingsMsg'
Learn More

Lovibond™ Replacement Reagent for CHECKIT™ Comparator

Lovibond™ Replacement Reagent for CHECKIT™ Comparator is manufactured under tightly controlled conditions to maintain the highest quality standards.

Brand:  Lovibond™ 461185

 View more versions of this product

Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.

Product Code. 10787833

  • 20.00 € / 15mL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Lovibond™ Replacement Reagent for CHECKIT™ Comparator is manufactured under tightly controlled conditions to maintain the highest quality standards.

  • Highest quality production standards
  • Consistent and reliable reagents are provided in liquid and tablet formats
  • Maximum accuracy
Specifications

Specifications

Yellow
0.1
Water analysis
15 mL
20°C
Nitric Acid 2.5 to 5%
Liquid
Colorimeter and spectrophotometer
0 to 0.5mg/L
DEHA Test Solution
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Lovibond™ Replacement Reagent for CHECKIT™ Comparator > DEHA solution; 15mL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.