Learn More
Lovibond™ Replacement Reagent for CHECKIT™ Comparator
Lovibond™ Replacement Reagent for CHECKIT™ Comparator is manufactured under tightly controlled conditions to maintain the highest quality standards.
20.00 € - 82.35 €
Specifications
For Use With (Application) | Water analysis |
---|---|
For Use With (Equipment) | Colorimeter and spectrophotometer |
Product Code | Brand | Format | Quantity | Type | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Format | Quantity | Type | Price | Quantity & Availability | ||||||
15392035
|
Lovibond™
461181 |
Liquid | 100 mL | DEHA Test Solution |
53.30 €
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
10787833
|
Lovibond™
461185 |
Liquid | 15 mL | DEHA Test Solution |
20.00 €
15mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
15353442
|
Lovibond™
512390BT |
Tablet | 100 Pack | EDTA Test Tablets |
44.40 €
100 tablets |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
15301995
|
Lovibond™
502171 |
Tablet | 250 Pack | DPD Glycine Test Tablets (for Cl separation) |
32.70 €
Pack of 250 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
12848121
|
Lovibond™
502171 |
Tablet | 250 Pack | DPD Glycine Test Tablets (for Cl separation) |
32.70 €
Each |
Estimated Shipment: 11-06-2025
Log in to see stock. |
||||||
15331745
|
Lovibond™
512171BT |
Tablet | 200 Pack | DPD Glycine Test Tablets |
82.35 €
Pack of 250 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
15368970
|
Lovibond™
512171BT |
Tablet | 200 Pack | DPD Glycine Test Tablets |
82.35 €
Pack of 250 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
15341845
|
Lovibond™
515440BT |
Tablet | 100 Pack | pH-Universal | N/A | N/A | N/A | |||||
Description
Lovibond™ Replacement Reagent for CHECKIT™ Comparator is manufactured under tightly controlled conditions to maintain the highest quality standards.
- Highest quality production standards
- Consistent and reliable reagents are provided in liquid and tablet formats
- Maximum accuracy
Specifications
Water analysis | |
Colorimeter and spectrophotometer |
Safety and Handling
- DEHA reagent solution
Signal Word
- Danger
Hazard Category
- Substances and mixtures corrosive to metals Category 1
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 2
Hazard Statement
- H290-May be corrosive to metals.
- H318-Causes serious eye damage.
- H315-Causes skin irritation.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P390-Absorb spillage to prevent material damage.
Supplemental information
- MIXTURE LIST-Contains : nitric acid
Your input is important to us. Please complete this form to provide feedback related to the content on this product.