Learn More
Lovibond™ Replacement Reagent for CHECKIT™ Comparator
Lovibond™ Replacement Reagent for CHECKIT™ Comparator is manufactured under tightly controlled conditions to maintain the highest quality standards.
Brand: Lovibond™ 502171
This item is not returnable.
View return policy
Description
Lovibond™ Replacement Reagent for CHECKIT™ Comparator is manufactured under tightly controlled conditions to maintain the highest quality standards.
- Highest quality production standards
- Consistent and reliable reagents are provided in liquid and tablet formats
- Maximum accuracy
Specifications
Tablet | |
Colorimeter and spectrophotometer | |
DPD Glycine Test Tablets (for Cl separation) |
Water analysis | |
250 Pack |
Product Suggestions
Customers who viewed this item also viewed
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Lovibond™ Replacement Reagent for CHECKIT™ Comparator > DPD Glycine Test Tablets (for Cl separation), 250/Pk.
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur