Learn More
Lovibond™ Replacement Reagent for CHECKIT™ Comparator
Lovibond™ Replacement Reagent for CHECKIT™ Comparator is manufactured under tightly controlled conditions to maintain the highest quality standards.
Brand: Lovibond™ 512390BT
Description
Lovibond™ Replacement Reagent for CHECKIT™ Comparator is manufactured under tightly controlled conditions to maintain the highest quality standards.
- Highest quality production standards
- Consistent and reliable reagents are provided in liquid and tablet formats
- Maximum accuracy
Specifications
White | |
4.5 | |
Water analysis | |
100 Pack | |
EDTA Test Tablets |
Boric Acid 40 to 45%, non-hazardous components 40 to 45%, EDTA Disodium Salt 5 to 15% | |
Tablet | |
Colorimeter and spectrophotometer | |
0.02 to 0.5mg/L |
Product Suggestions
Customers who viewed this item also viewed.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.