Learn More
BRAND™ Low Form Borosilicate Glass Beakers
Borosilicate glass construction. Squat form offers stability low risk of spillage.
78.95 € - 327.00 €
Specifications
Material Borosilicate Glass
Includes Integrated Spout
Graduated Yes
Certifications/Compliance ISO 3819, DIN 12331
Style Low Form
View More Specs
Products 10
Product Code Brand Capacity (Metric) Price Quantity & Availability  
Product Code Brand Capacity (Metric) Price Quantity & Availability  
10622822
View Documents
BRAND™
91253
800 mL
234.00 €
Pack of 10
In Stock
to see stock.
Add to basket
10411063
View Documents
BRAND™
91236
250 mL
121.00 €
Pack of 10
In Stock
to see stock.
Edge
Add to basket
10517461
View Documents
BRAND™
91254
1000 mL
256.00 €
Pack of 10
In Stock
to see stock.
Edge
Add to basket
10059920
View Documents
BRAND™
91268
3000 mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
30130370
View Documents
BRAND™
91268
3000 mL
327.00 €
Pack of 4
10142061
View Documents
BRAND™
91214
25 mL
119.00 €
Pack of 10
In Stock
to see stock.
Add to basket
10213731
View Documents
BRAND™
91248
600 mL
172.00 €
Pack of 10
In Stock
to see stock.
Add to basket
10345041
View Documents
BRAND™
91241
400 mL
156.00 €
Pack of 10
In Stock
to see stock.
Edge
Add to basket
10748241
View Documents
BRAND™
91224
100 mL
109.00 €
Pack of 10
In Stock
to see stock.
Edge
Add to basket
10090091
View Documents
BRAND™
91229
150 mL
117.00 €
Pack of 10
In Stock
to see stock.
Edge
Add to basket
Description
Description
  • Borisilicate glass construction (Boro 3.3)
  • Conforms to DIN 12331 / ISO 3819 standards
  • Includes graduations in white
  • Integrated spout provides easy, spill-proof pouring
Specifications
Specifications
Borosilicate Glass
Yes
Low Form
Integrated Spout
ISO 3819, DIN 12331