Learn More
Buerkle™ Lowering Cable for Buerkle™ Dipping Bottles
Buerkle™ Lowering Cable for dipping bottles and immersion cylinders, matching manually-operated drum reels made of various materials.
Brand:  Buerkle™ 5309-0150
259.12 EUR valid until 2024-12-31
Use promo code "23894" to get your promotional price.
 View more versions of this product
Product Code. 10381132
  • 303.00 € / Each
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Good chemical resistance
  • For dipping bottles and immersion cylinders, matching manually-operated drum reels made of various materials
  • Tensile load max. up to 10kg
Specifications
Specifications
Lowering Cable
PTFE-coated
2 mm
Stainless Steel (V2A)
Dipping Samplers and Immersion Cylinders, Manually-Operated Drum Reels
50 m