Learn More
Buerkle™ Lowering Cable for Buerkle™ Dipping Bottles
Buerkle™ Lowering Cable for dipping bottles and immersion cylinders, matching manually-operated drum reels made of various materials.
17.55 € - 687.00 €
Specifications
Product Type Lowering Cable
View More Specs
Products 15
Product Code Brand Material Length (Metric) Diameter (Metric) Outer Price Quantity & Availability  
Product Code Brand Material Length (Metric) Diameter (Metric) Outer Price Quantity & Availability  
11882663
missing translation for 'viewDocuments'
Buerkle™
5309-0410
Copper 10 m 4.5 mm
154.00 €
Each
In Stock
to see stock.
Add to basket
11892663
missing translation for 'viewDocuments'
Buerkle™
5309-0425
Copper 25 m 4.5 mm
342.00 €
Each
In Stock
to see stock.
Add to basket
11802673
missing translation for 'viewDocuments'
Buerkle™
5309-0450
Copper 50 m 4.5 mm
662.00 €
Each
In Stock
to see stock.
Add to basket
10066361
missing translation for 'viewDocuments'
Buerkle™
5309-0205
Polypropylene 50 m 2 mm
76.75 €
Each
In Stock
to see stock.
Add to basket
10157871
missing translation for 'viewDocuments'
Buerkle™
5309-0202
Polypropylene 25 m 2 mm
39.85 €
Each
In Stock
to see stock.
Add to basket
10279771
missing translation for 'viewDocuments'
Buerkle™
5309-0201
Polypropylene 10 m 2 mm
17.55 €
Each
In Stock
to see stock.
Add to basket
10107781
missing translation for 'viewDocuments'
Buerkle™
5309-0301
Stainless Steel (V2A) 10 m 1.25 mm
44.80 €
Each
In Stock
to see stock.
Add to basket
10498421
missing translation for 'viewDocuments'
Buerkle™
5309-0302
Stainless Steel (V2A) 25 m 1.25 mm
93.70 €
Each
In Stock
to see stock.
Add to basket
10084800
missing translation for 'viewDocuments'
Buerkle™
5309-0110
Stainless Steel (V2A) 10 m 2 mm
65.65 €
Each
In Stock
to see stock.
Add to basket
10779972
missing translation for 'viewDocuments'
Buerkle™
5309-0125
Stainless Steel (V2A) 25 m 2 mm
157.00 €
Each
In Stock
to see stock.
Add to basket
10408431
missing translation for 'viewDocuments'
Buerkle™
5309-0305
Stainless Steel (V2A) 50 m 1.25 mm
174.00 €
Each
In Stock
to see stock.
Add to basket
10381132
missing translation for 'viewDocuments'
Buerkle™
5309-0150
Stainless Steel (V2A) 50 m 2 mm
303.00 €
Each
In Stock
to see stock.
Add to basket
17270574
missing translation for 'viewDocuments'
Buerkle™
5309-0525
25 m
391.00 €
Each
In Stock
to see stock.
Add to basket
10579902
missing translation for 'viewDocuments'
Buerkle™
5309-0510
Stainless Steel 10 m 2.3 mm
166.00 €
Each
In Stock
to see stock.
Add to basket
17280574
missing translation for 'viewDocuments'
Buerkle™
5309-0550
100 m
687.00 €
Each
In Stock
to see stock.
Add to basket
Description
Description
  • Good chemical resistance
  • For dipping bottles and immersion cylinders, matching manually-operated drum reels made of various materials
  • Tensile load max. up to 10kg
Specifications
Specifications
Lowering Cable