missing translation for 'onlineSavingsMsg'
Learn More

RL Enterprises™ Natural Gas Micro Burner with NF Screwing G 1/2 Connector

A nickel-plated brass burner with air regulation.

Brand:  RL Enterprises™ 4014/1BSP

Product Code. 11735406

  • 38.10 € / Each
Estimated Shipment: 08-07-2025
to see stock.
Explore more special offers
Add to basket

Description

Description

  • Temperature range from 900° to about 1000°C
  • Without pilot light
  • Gas regulation by needle valve
  • Chrome steel burner, cast iron base
Specifications

Specifications

Micro Burner
3 mm
88 mm
1 Ea.
Butane, Propane, LPG
NF Screwing G 1/2 Connector
with NF Screwing G 1/2 Connector
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
RL Enterprises™ Natural Gas Micro Burner with NF Screwing G 1/2 Connector > Height: 88mm

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.