Learn More
STEMCELL Technologies Lymphoprep™
Density gradient medium for the isolation of mononuclear cells
Brand:  STEMCELL Technologies 07801
Encompass_Preferred
 View more versions of this product
Product Code. 17171036
  • 168.75 € / 250mL
Estimated Shipment: 10-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Lymphoprep™ is a density gradient medium recommended for the isolation of mononuclear cells from peripheral blood, cord blood, and bone marrow by exploiting differences in cell density
  • Granulocytes and erythrocytes have a higher density than mononuclear cells and therefore sediment through the Lymphoprep™ layer during centrifugation
  • Lymphoprep™ can be substituted for Ficoll-Paque™ without any need to change your existing protocols and is fully compatible with both SepMate™ and RosetteSep™
  • Lymphoprep™ has a density of 1.077 g/mL
Specifications
Specifications
250 mL
Mononuclear Cells
Cell Isolation