Learn More
Thermo Scientific™ M10 Microplate Swinging Bucket Rotor for Sorvall ST8/ST8R, Megafuge 8/8R and SL8/SL8R Centrifuges
Spin up to 4 microplates or 2 deepwell plates with the convenience of transportable microplate carriers.
Brand: Thermo Scientific™ 75005706
1337.53 EUR valid until 2025-04-30
Use promo code "24119" to get your promotional price.
Description
Ideally suited for quick spins—centrifuge 4 microplates or 2 midi-deepwell plates with the convenience of transportable carriers using the Thermo Scientific™ M10 Microplate Swinging Bucket Rotor.
Spin up to 4 standard or 2 midi-deepwell microplates with optional ClickSeal biocontainment lids, certified by Public Health England, Porton Down, UK.
Non-shielded design provides quick access to microplates.
- Secure, push-button Auto-Lock rotor exchange in less than 3 seconds delivers:
- Trouble-free rotor installation and removal
- No tools are required
- No complicated rotor changing procedures
- Clear chamber access for cleaning convenience
- Quick switch between applications, from conical tubes to blood tubes, from microtubes to microplates, and the flexibility to evolve with the changing needs of your laboratory
Ordering Information:
Unsealed buckets (75005723) and sealed buckets (75005721) are sold separately. Contact your sales representative for more information.
Specifications
M10 Microplate Swinging Bucket Rotor | |
4 Standard or 2 Deepwell Microplates | |
4,400 rpm | |
13103 | |
90° |
Swinging Bucket Rotor | |
2,576 x g | |
119 mm | |
Thermo Scientific Sorvall ST 8, Megafuge 8 and SL 8 Centrifuges | |
Yes |