missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ MAbPac™ Reversed Phase HPLC Columns

Use the new Thermo Scientific™ MAbPac™ RP columns for high-performance reversed-phase (RP) HPLC and LC/MS characterization of monoclonal antibodies (mAbs), fragments, variants, antibody drug conjugates (ADCs), PEGylated proteins, and bispecific proteins.

Brand:  Thermo Scientific™ 303184

1163.42 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.


 View more versions of this product

Product Code. 16874106

  • 1169.24 € / Each
Estimated Shipment: 08-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Use the new Thermo Scientific™ MAbPac™ Reversed Phase columns for high-performance reversed-phase (RP) HPLC and LC/MS characterization of monoclonal antibodies (mAbs), fragments, variants, antibody drug conjugates (ADCs), PEGylated proteins, and bispecific proteins. These columns have excellent resolution, long column lifetime, and low carry-over. The surface chemistry, particle size, and pore size, have been optimized for high resolution separations.

Compatible with organic solvent used in HPLC and MS under RP conditions
  • Resin is based on a hydrophobic supermacroporous 4μm polymer particles
  • Stable at extreme pH (0 to 14) and high temperature (up to 110 °C)
Efficient with low carry over
  • 1,500Å large pore size polymeric resin
  • Efficient separation of protein molecules with very low carry over
Designed for reversed-phase biomolecule LC and LC/MS separations
  • Superior resolution power for monoclonal antibodies and related substances
  • High efficiency with low carry-over
  • Excellent MS compatibility
  • Wide operating pH range: 0 to 14
  • High temperature stability: up to 110°C
  • High throughput
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Analytical Column
75 to 150 μL/min
4000 psi (275 bar)
1 to 7
Each
PSDVB (Polystyrene Divinylbenzene)
PSDVB (Polystyrene Divinylbenzene)
Reversed Phase
1 mm
150 mm
4 μm
1500 Å
110°C
MAbPac
Reversed Phase
Bio-LC
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.