missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ MAbPac™ Reversed Phase HPLC Columns

Use the new Thermo Scientific™ MAbPac™ RP columns for high-performance reversed-phase (RP) HPLC and LC/MS characterization of monoclonal antibodies (mAbs), fragments, variants, antibody drug conjugates (ADCs), PEGylated proteins, and bispecific proteins.

238.55 € - 1169.24 €

Specifications

Max. Pressure 4000 psi (275 bar)
Particle Size 4 μm
Pore Size 1500 Å
Phase Reversed Phase
View More Specs

Products 11
Product Code Brand Length (Metric) Diameter (Metric) Stationary Phase Price Quantity & Availability  
Product Code Brand Length (Metric) Diameter (Metric) Stationary Phase Price Quantity & Availability  
15407162
View Documents
Thermo Scientific™
088644
100 mm 3 mm Phenyl (Ph)
925.53 €
Each
Estimated Shipment: 14-03-2025
to see stock.
Add to basket
 
15417162
View Documents
Thermo Scientific™
088645
50 mm 3 mm Phenyl (Ph)
791.39 €
Each
Estimated Shipment: 10-03-2025
to see stock.
Add to basket
 
15427162
View Documents
Thermo Scientific™
088646
10 mm 3 mm Phenyl (Ph)
238.55 €
Pack of 2
Estimated Shipment: 15-01-2025
to see stock.
Add to basket
 
15308539
View Documents
Thermo Scientific™
088647/CPQ-00713550
100 mm 2.1 mm Phenyl (Ph)
929.63 €
Each
In Stock
to see stock.
Add to basket
 
15437162
View Documents
Thermo Scientific™
088648
50 mm 2.1 mm Phenyl (Ph)
791.39 €
Each
In Stock
to see stock.
Edge
Add to basket
 
15447162
View Documents
Thermo Scientific™
088649
10 mm 2.1 mm Phenyl (Ph)
238.55 €
Pack of 2
Estimated Shipment: 16-01-2025
to see stock.
Add to basket
 
15776222
View Documents
Thermo Scientific™
302597
50 mm 1 mm Phenyl (Ph) This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
16853740
View Documents
Thermo Scientific™
303182
50 mm 1 mm PSDVB (Polystyrene Divinylbenzene)
995.69 €
Each
In Stock
to see stock.
Add to basket
 
16864106
View Documents
Thermo Scientific™
303183
100 mm 1 mm PSDVB (Polystyrene Divinylbenzene)
1052.31 €
Each
Estimated Shipment: 03-01-2025
to see stock.
Add to basket
 
16874106
View Documents
Thermo Scientific™
303184
150 mm 1 mm PSDVB (Polystyrene Divinylbenzene)
1169.24 €
Each
Estimated Shipment: 03-01-2025
to see stock.
Add to basket
 
16307045
View Documents
Thermo Scientific™
303270
150 mm 2.1 mm
1000.62 €
Each
Estimated Shipment: 12-03-2025
to see stock.
Add to basket
 
Description

Description

Use the new Thermo Scientific™ MAbPac™ Reversed Phase columns for high-performance reversed-phase (RP) HPLC and LC/MS characterization of monoclonal antibodies (mAbs), fragments, variants, antibody drug conjugates (ADCs), PEGylated proteins, and bispecific proteins. These columns have excellent resolution, long column lifetime, and low carry-over. The surface chemistry, particle size, and pore size, have been optimized for high resolution separations.

Compatible with organic solvent used in HPLC and MS under RP conditions

  • Resin is based on a hydrophobic supermacroporous 4μm polymer particles
  • Stable at extreme pH (0 to 14) and high temperature (up to 110 °C)

Efficient with low carry over

  • 1,500Å large pore size polymeric resin
  • Efficient separation of protein molecules with very low carry over

Designed for reversed-phase biomolecule LC and LC/MS separations

  • Superior resolution power for monoclonal antibodies and related substances
  • High efficiency with low carry-over
  • Excellent MS compatibility
  • Wide operating pH range: 0 to 14
  • High temperature stability: up to 110°C
  • High throughput
Specifications

Specifications

4000 psi (275 bar)
4 μm
1500 Å
Reversed Phase
Videos
SDS
Documents

Documents

Product Certifications

Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.