missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva MabSelect™ Chromatography Media
Protein A affinity medium designed for capturing MAbs from large sample volumes
1093.00 € - 4367.00 €
Specifications
For Use With (Application) | For Industrial AppliCatIons and Well-Established in Approved Processes |
---|---|
Storage Requirements | 2°C to 8°C |
Binding Capacity | 30 mg Human lgG/mL (at 2.4 min. residence time) |
Product Type | MabSelect Media |
Flow Rate | <500 cm/Hoμr |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11339981
|
Cytiva
17-5199-01 |
25 mL |
1093.00 €
25mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11359981
|
Cytiva
17-5199-02 |
200 mL |
4367.00 €
200mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- High-flow agarose matrix allows for high flow velocities at production scale
- BioProcess medium supported for industrial applications and well-established in approved processes
- Low non-specific binding resulting in low levels of impurities in the product eluate pool
- Oriented coupling of ligand and optimized matrix gives high dynamic binding capacity
- Based on highly cross-linked agarose matrix with recombinant protein A ligand
- Ligand is engineered to favor oriented coupling and expressed in E. coli and is free of components of mammalian origin
- Hydrophilic nature of the base matrix ensures low levels of non-specific
- Average particle size: 85μm (d50V)
- pH Stability cleaning-in-place (CIP): 2 to 12
- pH Stability working range: 3 to 10
- Ligand: recombinant protein A (E.coli)
- Matrix: highly cross-linked agarose
- Binding capacity/mL chromatography medium: ∽30mg human lgG/mL at 2.4 min. residence time
- Pressure/Flow specification: <500cm/hr., <200 kPa, BPG 300, bed height 20cm
Specifications
For Industrial AppliCatIons and Well-Established in Approved Processes | |
30 mg Human lgG/mL (at 2.4 min. residence time) | |
<500 cm/Hoμr | |
7.87 in. | |
Certificate of Analysis | |
Highly Cross-Linked Agarose | |
Recombinant Protein A (E. coli) |
2°C to 8°C | |
MabSelect Media | |
85 μm | |
20 cm | |
<200 kPa | |
3 to 10 |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- MabSelect, 25 ml
Signal Word
- Warning
Hazard Category
- Flammable liquid Category 3
Hazard Statement
- H226-Flammable liquid and vapour.
Precautionary Statement
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : ethanol
- UNKNOWN ACUTE-16.5 per cent of the mixture consists of component(s) of unknown acute toxicity.
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Cytiva MabSelect™ Chromatography Media