missing translation for 'onlineSavingsMsg'
Learn More

Cole-Parmer™ EH-200 Series Macro-Kjeldahl Extraction Heater

The EH-200 Series Macro-Kjeldahl Extraction Heater is designed to determine the protein content of food on a macro scale, from 100 to 800 mL.

Brand:  Cole-Parmer™ 03075-22

 View more versions of this product

Product Code. 11844381

  • 4725.00 € / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Independent heat controller lets you adjust the temperature range of each mantle individually.
  • Stainless steel construction is rugged and easy to clean.
  • Run several projects simultaneously using these multiposition macro-kjeldahl extraction heaters.
  • Heaters are available with two or six individually controlled heating mantles that accommodate either 100 to 300 mL vessels or 500 to 800 mL vessels.
  • Back-mounted brackets hold flask support rods up to 12.7 mm in diameter.
  • Each mantle has an independent heater with control dial, on/off switch, and indicator light, and operates to a maximum element temperature of 800°C.
  • A quartz heating element is housed in a thermally insulated cartridge for easy replacement, extending the life of the heater.
  • Heat controllers located in the lower part of the unit is safely separated from the upper heating elements by a stainless steel tray and well-ventilated air space.
  • Extra safety features include an earth screen and double fuses on the supply line.
Specifications

Specifications

500 to 800 mL (16.9 to 27.1 oz.)/well
260 x 970 x 165 mm
165 mm
230 V
Ceramic Fiber/Mineral Wool
3300 W
5.8 kg
38.2 in.
Extraction Heater/Stirrer
16.9 to 27.1 oz.
500 to 800 mL/well
6.5 in.
260 mm
Stainless steel
UK Plug, Wired Directly
12.8 lb.
6
970 mm
800°C
50/60 Hz
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Cole-Parmer™ EH-200 Series Macro-Kjeldahl Extraction Heater > EH-200 Series Macro-Kjeldahl Extraction Heater; Capacity: 500 to 800 mL (16.9 to 27.1 oz.)/well; Well Count: 6; Wattage: 3300 W; Power Supply: UK Plug, Wired Directly

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.